Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4 Rat anti-Mouse, Alexa Fluor™ 594, Clone: MTS510, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP224865AF594

Catalog No. NP22486A94


Add to cart

Description

Description

TLR4 Monoclonal antibody specifically detects TLR4 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

TLR4
MTS510
Flow Cytometry, Flow (Cell Surface)
Conjugated, Purified
TLR4
This antibody was developed by immunizing rats with the Ba/F3 cell line expressing mouse TLR4 and MD-2 (Akashi et al, 2000).
Protein G purified
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary
7099.0
Mouse
Flow Cytometry
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
Rat
IgG2a Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
In FA, the antibody blocks activation of monocytes with LPS (Akashi et al. 2000). This antibody preferentially recognizes TLR4-MD-2 complex than of TLR4 alone. The optimal condition has to be determined for individual experiments.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit