Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR4 Rat anti-Mouse, Alexa Fluor™ 750, Clone: MTS510, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP224865AF750
Description
TLR4 Monoclonal specifically detects TLR4 in Mouse samples. It is validated for Flow Cytometry, Flow (Cell Surface).Specifications
TLR4 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
TLR4 | |
This antibody was developed by immunizing rats with the Ba/F3 cell line expressing mouse TLR4 and MD-2 (Akashi et al, 2000). | |
0.1 mL | |
Primary | |
In FA, the antibody blocks activation of monocytes with LPS (Akashi et al. 2000). This antibody preferentially recognizes TLR4-MD-2 complex than of TLR4 alone. The optimal condition has to be determined for individual experiments. | |
Store at 4°C in the dark. |
Flow Cytometry | |
MTS510 | |
Flow Cytometry, Flow (Cell Surface) | |
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
Rat | |
Protein G purified | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
7099.0 | |
Mouse | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title