Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR5 Mouse anti-Human, Alexa Fluor 405, Clone: 624915, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6704V100UG
This item is not returnable.
View return policy
Description
TLR5 Monoclonal specifically detects TLR5 in Human samples. It is validated for Flow Cytometry.Specifications
TLR5 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O60602 | |
TLR5 | |
E. coli-derived recombinant human TLR5 Ile21-Phe115 Accession # O60602 | |
RUO | |
7100 | |
Human | |
IgG1 |
Flow Cytometry | |
624915 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
MGC126430, MGC126431, SLEB1FLJ10052, TIL-3, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
Mouse | |
100 μg | |
Primary | |
Detects human TLR5 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) TLR1, 2, 3, 4, 7, 8, recombinant mouse TLR5, 6, or rhTIRAP is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)