Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR9 Mouse anti-Mouse, Unconjugated, Clone: NaR9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABF2156100UL
Description
TLR9 Monoclonal antibody specifically detects TLR9 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Inhibits Activity/Function.Specifications
TLR9 | |
Monoclonal | |
Unconjugated | |
Toll-like receptor 9;CD289 | |
Mouse | |
116.41 kDa calculated. | |
100 μL | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunocytochemistry | |
NaR9 | |
NP_112455.2 | |
Tlr9 | |
Ba/F3 cells expressing full-length mouse TLR9 protein. Immunization was carried out on Tlr9 -/- mice. | |
Protein G Purified | |
Inflammation and Immunology | |
Clone NaR9 is a mouse monoclonal antibody that specifically detects Toll-like receptor 9 in murine species. It targets an epitope within the N-terminal region. | |
Stable for 1 year at -20°C from date of receipt. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction