Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR9 Mouse anti-Mouse, Unconjugated, Clone: NaR9, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen TLR9
Applications Flow Cytometry, Immunocytochemistry
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF21561MI
View Documents
MilliporeSigma
MABF2156100UL
100 μL
Each for $372.00
Add to cart
 
MABF21562MI
View Documents
MilliporeSigma
MABF215625UL
25 μL
Each for $147.00
Add to cart
 
Description

Description

TLR9 Monoclonal antibody specifically detects TLR9 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Inhibits Activity/Function.
Specifications

Specifications

TLR9
Unconjugated
Mouse
Toll-like receptor 9;CD289
Ba/F3 cells expressing full-length mouse TLR9 protein. Immunization was carried out on Tlr9 -/- mice.
Primary
Stable for 1 year at -20°C from date of receipt.
Flow Cytometry, Immunocytochemistry
Purified
Mouse
Tlr9
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit