Learn More
TLR9 Rabbit anti-Mouse, Alexa Fluor 350, Clone: 1138D, R&D Systems™
Rabbit MonoclonalAntibody
Manufacturer:  R&D Systems FAB7960U100UG
Catalog No. FAB7960U100
Add to cart
This item is not returnable. View return policy
Description
Description
Recombinant Monoclonal Antibody.
Specifications
Specifications
TLR9
Monoclonal
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
CD289, CD289 antigen, toll-like receptor 9
Chinese hamster ovary cell line CHO-derived recombinant mouse TLR9 Leu26-Asp818 Accession # AAK29625
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse TLR-9 in direct ELISAs.
Flow Cytometry
1138D
Flow Cytometry 0.25-1 ug/10^6 cells
AAK29625
Rabbit
IgG
RUO
Primary
54106
Mouse
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)