Learn More
Brady™ TLS 2200™ Series Printer Ribbon - R8600 Resin, White
This self contained super-tough ribbon cartridge for the TLS 2200™ and TLS-PC Link™ Thermal Labeling Systems ensures your printed text lasts for years in industrial areas.
Manufacturer:  Brady™ R6810WT
Catalog No. 19-104-663
Add to cart
Description
Description
  • One-color ribbon for printing a single color onto a label
  • R6800 Series white resin ribbons offer excellent solvent, scratch and smudge resistance when printed on the appropriate label material from Brady™
  • Recommended for printing on dark-colored heat shrinkable tubing materials, such as black B-345 PermaSleeve™ High Temperature Wire Markers from Brady™
  • For use with TLS 2200™ and TLS-PC Link™ Thermal Labeling Systems (not compatible with the BMP™71 Label Printer)
Specifications
Specifications
Ribbon in. Printer Specific Cartridge
White
TLS-PC LINK™ Thermal Labeling System;TLS 2200™ Thermal Labeling System
Roll of 65 Feet
Rectangle
Printer Ribbon
Not Autoclavable
Printer Ribbon
65 ft.
Roll of 65 Feet
113.03 mm
Roll of 65 Feet