Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM2D2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP17984520UL

 View more versions of this product

Catalog No. NBP17984520


Add to Cart

Description

Description

TM2D2 Polyclonal specifically detects TM2D2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TM2D2
Polyclonal
Western Blot 1:1000
NP_510882
TM2D2
Synthetic peptide directed towards the middle region of human TM2D2The immunogen for this antibody is TM2D2. Peptide sequence QELGYGCLKFGGQAYSDVEHTSVQCHALDGIECASPRTFLRENKPCIKYT.
Affinity Purified
RUO
83877
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
BBP-like protein 1, Beta-amyloid-binding protein-like protein 1, BLP1MGC125814, MGC125813, TM2 domain containing 2, TM2 domain-containing protein 2
Rabbit
23 kDa
20 μL
Primary
Human
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit