Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TM2D2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179845
Description
TM2D2 Polyclonal specifically detects TM2D2 in Human samples. It is validated for Western Blot.Specifications
TM2D2 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
NP_510882 | |
TM2D2 | |
Synthetic peptide directed towards the middle region of human TM2D2The immunogen for this antibody is TM2D2. Peptide sequence QELGYGCLKFGGQAYSDVEHTSVQCHALDGIECASPRTFLRENKPCIKYT. | |
Affinity Purified | |
RUO | |
83877 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
BBP-like protein 1, Beta-amyloid-binding protein-like protein 1, BLP1MGC125814, MGC125813, TM2 domain containing 2, TM2 domain-containing protein 2 | |
Rabbit | |
23 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Zebrafish: 77% Chicken: 76%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title