Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TM2D2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | TM2D2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17984520
|
Novus Biologicals
NBP17984520UL |
20 μL |
Each for $152.22
|
|
NBP179845
|
Novus Biologicals
NBP179845 |
100 μL |
Each for $436.00
|
|
Description
TM2D2 Polyclonal specifically detects TM2D2 in Human samples. It is validated for Western Blot.Specifications
TM2D2 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
BBP-like protein 1, Beta-amyloid-binding protein-like protein 1, BLP1MGC125814, MGC125813, TM2 domain containing 2, TM2 domain-containing protein 2 | |
TM2D2 | |
IgG | |
Affinity Purified | |
23 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_510882 | |
83877 | |
Synthetic peptide directed towards the middle region of human TM2D2The immunogen for this antibody is TM2D2. Peptide sequence QELGYGCLKFGGQAYSDVEHTSVQCHALDGIECASPRTFLRENKPCIKYT. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title