Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM4SF1/L6 Mouse anti-Human, Alexa Fluor 405, Clone: 877621, R&D Systems™

Mouse MonoclonalAntibody

Manufacturer:  R&D Systems FAB8164V100UG

Catalog No. FAB8164V100


Add to cart
This item is not returnable. View return policy

Description

Description

TM4SF1/L6 Monoclonal antibody specifically detects TM4SF1/L6 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TM4SF1/L6
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P30408
TM4SF1
NS0 mouse myeloma cell line transfected with human TM4SF1/L6 Met1-Cys202 Accession # P30408
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human TM4SF1/L6 in ELISA. In Flow Cytometry, it stains HEK293 cells transfected with human TM4SF1/L6, and does not stain HEK293 cells transfected with irrelevant human cell surface marker.
Flow Cytometry
877621
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
L6, M3s1, M3S1Tumor-associated antigen L6, Membrane component chromosome 3 surface marker 1, membrane component, chromosome 3, surface marker 1, TAAL6, TAAL6H-L6, transmembrane 4 L six family member 1, transmembrane 4 L6 family member 1, transmembrane 4 superfamily member 1
Mouse
IgG1
RUO
Primary
4071
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)