Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TM4SF1/L6 Alexa Fluor 750-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8164S100UG
This item is not returnable.
View return policy
Description
TM4SF1/L6 Monoclonal specifically detects TM4SF1/L6 in Human samples. It is validated for Flow Cytometry.Specifications
TM4SF1/L6 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P30408 | |
TM4SF1 | |
NS0 mouse myeloma cell line transfected with human TM4SF1/L6 Met1-Cys202 Accession # P30408 | |
RUO | |
4071 | |
Human | |
IgG1 |
Flow Cytometry | |
877621 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
L6, M3s1, M3S1Tumor-associated antigen L6, Membrane component chromosome 3 surface marker 1, membrane component, chromosome 3, surface marker 1, TAAL6, TAAL6H-L6, transmembrane 4 L six family member 1, transmembrane 4 L6 family member 1, transmembrane 4 superfamily member 1 | |
Mouse | |
100 μg | |
Primary | |
Detects human TM4SF1/L6 in ELISA. In Flow Cytometry, it stains HEK293 cells transfected with human TM4SF1/L6, and does not stain HEK293 cells transfected with irrelevant human cell surface marker. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction