Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM4SF4 Mouse anti-Human, Alexa Fluor 350, Clone: 832441, R&D Systems™

Mouse MonoclonalAntibody

Manufacturer:  R&D Systems FAB7998U100UG

Catalog No. FAB7998U100


Add to cart
This item is not returnable. View return policy

Description

Description

TM4SF4 Monoclonal antibody specifically detects TM4SF4 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TM4SF4
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P48230
TM4SF4
NS0 mouse myeloma cell line transfected with human TM4SF4 Accession # P48230
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human TM4SF4 in ELISAs.
Flow Cytometry
832441
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
FLJ31015, il-TMP, ILTMP, ILTMPintestinal and liver (il) tetraspan membrane protein, Intestine and liver tetraspan membrane protein, transmembrane 4 L six family member 4, transmembrane 4 L6 family member 4, transmembrane 4 superfamily member 4
Mouse
IgG1
RUO
Primary
7104
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)