Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TM7SF4 Polyclonal Antibody
TM7SF4 Polyclonal Antibody
Supplier: Thermo Scientific PA569651
Description
TM7SF4 Polyclonal Antibody for Western Blot

Specifications
TM7SF4 | |
Polyclonal | |
Unconjugated | |
DCSTAMP | |
4833414I07Rik; DC-specific transmembrane protein; Dcstamp; DC-STAMP; Dendritic cells (DC)-specific transmembrane protein; Dendritic cell-specific transmembrane protein; dendrocyte expressed seven transmembrane protein; dendrocyte-expressed seven transmembrane protein; dentrocyte expressed seven transmembrane protein; FIND; hDC-STAMP; IL-4-induced protein; IL-Four (IL-4) induced; IL-Four INDuced; IL-four-induced protein; LOW QUALITY PROTEIN: dendritic cell-specific transmembrane protein; mDC-STAMP; TM7SF4; transmembrane 7 superfamily member 4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
81501 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H295 | |
DCSTAMP | |
synthetic peptide directed towards the N terminal region of human TM7SF4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction