Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM9SF2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548517

Catalog No. PA548517


Add to cart

Description

Description

In the intracellular compartments, may function as a channel or small molecule transporter.
Specifications

Specifications

TM9SF2
Polyclonal
PBS with 0.09% sodium azide
Q99805
Transmembrane 9 superfamily member 2, p76, TM9SF2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9375
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TM9SF2
Liquid
TM9SF2
KLH conjugated synthetic peptide between 111-140 amino acids from the N-terminal region of human TM9SF2.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit