Learn More
TMBIM6 Mouse anti-Human, Clone: 20F565, Abnova™
Mouse monoclonal antibody raised against human TMBIM6.
Manufacturer:  Abnova Corporation MAB12711
Catalog No. 89-935-019
Add to cart
Description
Description
Specifications
Specifications
TMBIM6
20F565
Mouse monoclonal antibody raised against human TMBIM6.
In PBS (0.05% BSA, 0.05% sodium azide).
BAXI1/BI-1/TEGT
Mouse
IgG2b, kappa
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
TMBIM6
A mixture of three synthetic peptides corresponding to the N-terminus, internal and C-terminus regions of human TMBIM6.
Protein G purification
RUO
Primary
7009