Learn More
Aviva Systems Biology Corp TMC8 Antibody - N-terminal region (ARP52991_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52991_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMC8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMC8. Alternate names: EV2, EVER2, EVIN2, MGC102701, MGC40121ApplicationWB. Species reactivity: Human
Catalog No. 50-014-63067