Tocris™ TMCB

Dual-kinase inhibitor; inhibits CK2 and ERK8

Manufacturer: Tocris™ 3675/50

 View more versions of this product

Catalog No. 36-755-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors TMCB
    Purity >98
    Molecular Weight 534.82
    Quantity 50mg
    Product Type TMCB

    Dual-kinase inhibitor; inhibits both casein kinase 2 (CK2) and extracellular-signal-regulated kinase 8 (ERK8) (IC50 = 0.50µM for both CK2 and ERK8). Displays selectivity for CK2 over protein kinases normally susceptible to CK2 inhibitors (Ki values are 0.25, 8.65, 11.90 and 15.25µM for CK2, PIM1, DYRK1a and HIPK2 respectively).

    • Chemical Name: 2-(4,5,6,7-Tetrabromo-2-(dimethylamino)-1H-benzo[d]imidazol-1-yl)acetic acid
    • Formula: C11H9Br4N3O2
    • CAS Number: 905105-89-7
    • Solubility: Soluble to 25mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit