Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMCC2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19156220UL

 View more versions of this product

Catalog No. NBP19156220


Add to cart

Description

Description

TMCC2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

TMCC2
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
TMCC2
Synthetic peptide directed towards the N terminal of human TMCC2. Peptide sequence GETTGANSAGGPTSDAGAAAAPNPGPRSKPPDLKKIQQLSEGSMFGHGLK.
Immunogen affinity purified
RUO
Primary
9911
Western Blot
Western Blot 1:1000
NP_055673
cerebral protein 11, FLJ38497, hucep-11, HUCEP11, KIAA0481, transmembrane and coiled-coil domain family 2, transmembrane and coiled-coil domains 2, transmembrane and coiled-coil domains protein 2
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit