Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMED3 Polyclonal Antibody
TMED3 Polyclonal Antibody
Supplier: Thermo Scientific PA542938
Description
TMED3 Polyclonal Antibody for Western Blot

Specifications
TMED3 | |
Polyclonal | |
Unconjugated | |
TMED3 | |
1200002G13Rik; AW546672; C15orf22; integral type I protein; membrane protein p24B; MGC133022; p24 family protein gamma-4; P24b; p24g4; p24gamma4; p26; testis tissue sperm-binding protein Li 48e; TMED3; transmembrane emp24 domain containing 3; transmembrane emp24 domain-containing protein 3; transmembrane emp24 domain-containing protein 3-like; transmembrane emp24 protein transport domain containing 3; transmembrane p24 trafficking protein 3; UNQ5357/PRO1078 | |
Rabbit | |
Protein A | |
RUO | |
23423 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y3Q3 | |
TMED3 | |
Synthetic peptide directed towards the C-terminal of human TMED3 (aa 168-217). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction