Learn More
TMED4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP2843030.1ML
Catalog No. NB032231
Add to cart
Description
Description
TMED4 Polyclonal specifically detects TMED4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
TMED4
Unconjugated
Purified
Rabbit
Protein A purified
RUO
Primary
222068
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
PBS, 2% Sucrose
Endoplasmic reticulum stress-response protein 25, endoplasmic reticulum stress-response protein 25 kDa, ERS25, HNLF, Putative NF-kappa-B-activating protein 156, putative NFkB activating protein HNLF, transmembrane emp24 domain-containing protein 4, transmembrane emp24 protein transport domain containing 4
The immunogen is a synthetic peptide directed towards the N-terminal region of human TMED4
0.1 mL
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Polyclonal
Human