Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMED6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548783

Catalog No. PA548783

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TMED6
Polyclonal
PBS with 0.09% sodium azide
Q8WW62
Transmembrane emp24 domain-containing protein 6, p24 family protein gamma-5, p24gamma5, TMED6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
146456
Western Blot
Unconjugated
TMED6
Liquid
TMED6
KLH conjugated synthetic peptide between 125-153 amino acids from the Central region of human TMED6.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit