Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMED8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548724

Catalog No. PA548724

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Specifications

Specifications

TMED8
Polyclonal
PBS with 0.09% sodium azide
Q6PL24
Protein TMED8, TMED8, FAM15B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
283578
Western Blot
Unconjugated
TMED8
Liquid
TMED8
KLH conjugated synthetic peptide between 154-182 amino acids from the Central region of human TMED8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit