Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEFF1/Tomoregulin-1 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0571-7S12, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211926AF350
Description
TMEFF1/Tomoregulin-1 Monoclonal specifically detects TMEFF1/Tomoregulin-1 in Human samples. It is validated for Western Blot.Specifications
TMEFF1/Tomoregulin-1 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
TMEFF1 | |
Human recombinant TMEFF1/Tomoregulin-1 | |
0.1 mL | |
Primary | |
Human | |
IgG2 |
Western Blot | |
MM0571-7S12 | |
Western Blot | |
C9orf2, H7365member 1, tomoregulin-1, TR-1, Transmembrane protein with EGF-like and one follistatin-like domain, transmembrane protein with EGF-like and two follistatin-like domains 1 | |
Mouse | |
Protein G purified | |
Core ESC Like Genes, Stem Cell Markers | |
8577.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title