Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM100 Mouse anti-Human, Clone: OTI10G3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524949

Catalog No. PIMA524949


Add to cart

Description

Description

Plays a role during embryonic arterial endothelium differentiation and vascular morphogenesis through the ACVRL1 receptor-dependent signaling pathway upon stimulation by bone morphogenetic proteins, such as GDF2/BMP9 and BMP10.
Specifications

Specifications

TMEM100
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NV29
Transmembrane protein 100, TMEM100
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55273
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI10G3
Unconjugated
TMEM100
Liquid
TMEM100
Full length human recombit protein of TMEM100 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit