Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM117 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 213141AP150UL
Description
TMEM117 is a protein coding gene.Specifications
TMEM117 | |
Polyclonal | |
Unconjugated | |
TMEM117 | |
TMEM117, transmembrane protein 117 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
320709, 84216 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BH18, Q9H0C3 | |
Tmem117 | |
TMEM117 Fusion Protein Ag15966 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction