Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tmem125 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549175

Catalog No. PA549175


Add to cart

Specifications

Specifications

Tmem125
Polyclonal
PBS with 0.09% sodium azide
Q8CHQ6
Transmembrane protein 125, Tmem125
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
230678
Western Blot
Unconjugated
Tmem125
Liquid
TMEM125
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of mouse Tmem125.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit