Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM132D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569683
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
TMEM132D may serve as a cell-surface marker for oligodendrocyte differentiation.
Specifications
TMEM132D | |
Polyclonal | |
Unconjugated | |
TMEM132D | |
C630028F04Rik; HBE120; hypothetical protein; KIAA1944; Mature OL transmembrane protein; mature oligodendrocyte transmembrane protein; mature oligodendrocytes transmembrane protein; MOLT; PPP1R153; protein phosphatase 1, regulatory subunit 153; TMEM132D; Transmembrane protein 132D | |
Rabbit | |
Affinity chromatography | |
RUO | |
243274 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q76HP3 | |
TMEM132D | |
synthetic peptide directed towards the middle region of human TMEM132D (aa 700-749). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction