Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM132E Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548214

Catalog No. PA548214

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Required for normal inner ear hair cell function and hearing.
Specifications

Specifications

TMEM132E
Polyclonal
PBS with 0.09% sodium azide
Q6IEE7, Q6IEE6
Transmembrane protein 132E, TMEM132E
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
124842, 270893
Flow Cytometry, Western Blot
Unconjugated
TMEM132E
Liquid
TMEM132E
KLH conjugated synthetic peptide between 704-733 amino acids from the C-terminal region of human TMEM132E.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit