Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM147 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282028AF700
This item is not returnable.
View return policy
Description
TMEM147 Polyclonal antibody specifically detects TMEM147 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
TMEM147 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 700 | |
MGC1936, NIFIE14, Protein NIFIE 14, seven transmembrane domain protein, transmembrane protein 147 | |
TMEM147 antibody was raised against a 13 amino acid synthetic peptide near the center of human TMEM147. The immunogen is located within amino acids 70 - 120 of TMEM147. Amino Acid Squence: AAMSRNAGKGEYKIM | |
0.1 mL | |
Cell Biology, Neuroscience | |
10430 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction