Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM14A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112745
Description
TMEM14A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Inhibits apoptosis via negative regulation of the mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway.
Specifications
TMEM14A | |
Polyclonal | |
Unconjugated | |
TMEM14A | |
C6orf73; PTD011; TMEM14A; Transmembrane protein 14A | |
Rabbit | |
Protein G | |
RUO | |
28978 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9Y6G1 | |
TMEM14A | |
Recombinant Human Transmembrane protein 14A protein (45-78AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction