Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM158 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549532

Catalog No. PA549532


Add to cart

Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

Receptor for brain injury-derived neurotrophic peptide (BINP), a synthetic 13-mer peptide.
Specifications

Specifications

TMEM158
Polyclonal
PBS with 0.09% sodium azide
Q8WZ71
Transmembrane protein 158, 40 kDa BINP-binding protein, p40BBP, Ras-induced senescence protein 1, TMEM158, HBBP, RIS1
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25907
Western Blot
Unconjugated
TMEM158
Liquid
TMEM158
KLH conjugated synthetic peptide between 186-218 amino acids from human TMEM158.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit