Learn More
Aviva Systems Biology Corp TMEM161A Antibody - middle region (ARP47355_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47355_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM161A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMEM161A. Alternate names: AROS-29, FLJ20422, FLJ39645ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-59521