Learn More
TMEM16A Rabbit anti-Human, KHL, Clone: 3J18, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB117125UL
 View more versions of this product
Catalog No. ZRB117125MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
TMEM16A Monoclonal antibody specifically detects TMEM16A in Human samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
TMEM16A
3J18
KHL
Purified
ANO1, DOG1, ORAOV2, TAOS2, TMEM16A
KLH-conjugated linear peptide corresponding to 15 amino acids from the extracellular C-terminal region of human Transmembrane protein 16A (TMEM16A).
Target molecular weight: 114.08 kDa calculated; ∼150 kDa observed. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_060513.5
Transmembrane protein 16A;Anoctamin-1;Discovered on gastrointestinal stromal tumors protein 1;Oral cancer overexpressed protein 2;Tumor-amplified and overexpressed sequence 2
Rabbit
IgG
Protein A Purified
Neuroscience
Primary
Human