Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM16A Rabbit anti-Human, KHL, Clone: 3J18, MilliporeSigma™

Rabbit Monoclonal Antibody

Manufacturer:  MilliporeSigma ZRB11714X25UL

 View more versions of this product

Catalog No. ZRB11714XMI

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

TMEM16A Monoclonal antibody specifically detects TMEM16A in Human samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

TMEM16A
3J18
KHL
Purified
ANO1, DOG1, ORAOV2, TAOS2, TMEM16A
KLH-conjugated linear peptide corresponding to 15 amino acids from the extracellular C-terminal region of human Transmembrane protein 16A (TMEM16A).
Target molecular weight: 114.08 kDa calculated; ∼150 kDa observed. Uncharacterized bands may be observed in some lysate(s).
4 x 25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_060513.5
Transmembrane protein 16A;Anoctamin-1;Discovered on gastrointestinal stromal tumors protein 1;Oral cancer overexpressed protein 2;Tumor-amplified and overexpressed sequence 2
Rabbit
IgG
Protein A Purified
Neuroscience
Primary
4X
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit