Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM16A Rabbit anti-Human, KHL, Clone: 3J18, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen TMEM16A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB117125MI
View Documents
MilliporeSigma
ZRB117125UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB11714XMI
View Documents
MilliporeSigma
ZRB11714X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

TMEM16A Monoclonal antibody specifically detects TMEM16A in Human samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

TMEM16A
KHL
Rabbit
Transmembrane protein 16A;Anoctamin-1;Discovered on gastrointestinal stromal tumors protein 1;Oral cancer overexpressed protein 2;Tumor-amplified and overexpressed sequence 2
KLH-conjugated linear peptide corresponding to 15 amino acids from the extracellular C-terminal region of human Transmembrane protein 16A (TMEM16A).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Purified
Human
ANO1, DOG1, ORAOV2, TAOS2, TMEM16A
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit