Learn More
Aviva Systems Biology Corp TMEM184A Antibody - C-terminal region (ARP54530_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54530_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM184A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TMEM184A. Alternate names: FLJ24011, MGC9712ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-61236