Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
TMEM195 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 213551AP
Catalog No. 50-173-3383
Description
Description
AGMO gene ontology annotations related to this gene include ether lipid metabolic process; lipid biosynthetic process; membrane lipid metabolic process.
Specifications
Specifications
TMEM195
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6ZNB7, Q8BS35
Alkylglycerol monooxygenase, TMEM195, transmembrane protein 195
Rabbit
IgG
150 μL
-20°C
Primary
319660, 392636
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
AGMO
Liquid
Agmo
TMEM195 Fusion Protein Ag15774
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse