Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM237 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598638
Description
TMEM237 Polyclonal Antibody for Western Blot, ELISA
The protein encoded by this gene is a tetraspanin protein that is thought to be involved in WNT signaling. Defects in this gene are a cause of Joubert syndrome-14. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jan 2012].
Specifications
TMEM237 | |
Polyclonal | |
Unconjugated | |
TMEM237 | |
AI853305; Als2cr4; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 4; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 4 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 4 protein homolog; Gm972; JBTS14; Tmem237; Transmembrane protein 237 | |
Rabbit | |
Protein G | |
RUO | |
65062 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3.33 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96Q45 | |
TMEM237 | |
Peptide sequence from Human Transmembrane protein 237 protein (59-78AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction