Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM255B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5118316

This item is not returnable.
View return policy
Description
Specificity: Human TMEM255B
TMEM255B Polyclonal antibody specifically detects TMEM255B in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
TMEM255B | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q8WV15 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
348013 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
TMEM255B | |
FAM70B; family with sequence similarity 70, member B; protein FAM70B; TMEM255B; Transmembrane protein 255B | |
E. coli-derived Human TMEM255B/FAM70B fragment | |
100 μL | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title