Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM30A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553193
Description
Immunogen sequence: VKSRDDSQLN GDSSALLNPS KECEPYRRNE DKPIAPCGAI ANSMFNDTLE LFLIGNDSYP IPIALKKKGI AWWTDKNVKF RNPPGGDNLE ERFKGTTKPV NWLKPVYMLD SDPDNNGFI Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 84%.
Accessory component of a P4-ATPase flippase complex which catalyzes the hydrolysis of ATP coupled to the transport of aminophospholipids from the outer to the inner leaflet of various membranes and ensures the maintenance of asymmetric distribution of phospholipids. Phospholipid translocation seems also to be implicated in vesicle formation and in uptake of lipid signaling molecules. The beta subunit may assist in binding of the phospholipid substrate. Required for the proper folding, assembly and ER to Golgi exit of the ATP8A2:TMEM30A flippase complex. ATP8A2:TMEM30A may be involved in regulation of neurite outgrowth, and, reconstituted to liposomes, predomiminantly transports phosphatidylserine (PS) and to a lesser extent phosphatidylethanolamine (PE). The ATP8A1:TMEM30A flippase complex seems to play a role in regulation of cell migration probably involving flippase-mediated translocation of phosphatidylethanolamine (PE) at the plasma membrane. Required for the formation of the ATP8A2, ATP8B1 and ATP8B2 P-type ATPAse intermediate phosphoenzymes. Involved in uptake of platelet-activating factor (PAF), synthetic drug alkylphospholipid edelfosine, and, probably in association with ATP8B1, of perifosine. Also mediates the export of alpha subunits ATP8A1, ATP8B1, ATP8B2, ATP8B4, ATP10A, ATP10B, ATP10D, ATP11A, ATP11B and ATP11C from the ER to other membrane localizations. [UniProt].
Specifications
TMEM30A | |
Polyclonal | |
Unconjugated | |
TMEM30A | |
2010200I23Rik; AW540225; C6orf67; Cdc50a; cell cycle control protein 50A; cell cycle control protein 50A; LOW QUALITY PROTEIN: cell cycle control protein 50A; Cell cycle control protein 50A-like protein; D9Wsu20e; FLJ10856; P4-ATPase flippase complex beta subunit TMEM30A; TMEM30A; Transmembrane protein 30A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55754 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NV96 | |
TMEM30A | |
Recombinant protein corresponding to Human TMEM30A. Recombinant protein control fragment (Product #RP-90931). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction