Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM33 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259137

Catalog No. NBP259137


Add to cart

Description

Description

TMEM33 Polyclonal antibody specifically detects TMEM33 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

TMEM33
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
DB83, FLJ10525, Protein DB83, SHINC3,1600019D15Rik, transmembrane protein 33
Rabbit
IgG
100 ul
Store at 4C.
Polyclonal
Human
Western Blot, Immunoprecipitation
Western Blot, Immunoprecipitation
Affinity Purified
TMEM33
Maps to a region between 197 and 247
Immunogen affinity purified
RUO
Primary
55161
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only