Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM38B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen TMEM38B
Immunogen Synthetic peptides corresponding to TMEM38B(transmembrane protein 38B) The peptide sequence was selected from the C terminal of TMEM38B. Peptide sequence WMLFGWQQPFSSCEKKSEAKSPSNGVGSLASKPVDVASDNVKKKHTKKNE.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15997620 View Documents Novus BiologicalsSupplier Diversity Partner
NBP15997620UL
20ul Each for $149.00
Add to cart
 
NBP159976 View Documents Novus BiologicalsSupplier Diversity Partner
NBP159976
100 ul Each for $329.00
Add to cart
 
Description

Description

TMEM38B Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

TMEM38B
Rabbit
Polyclonal
Unconjugated
55151
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to TMEM38B(transmembrane protein 38B) The peptide sequence was selected from the C terminal of TMEM38B. Peptide sequence WMLFGWQQPFSSCEKKSEAKSPSNGVGSLASKPVDVASDNVKKKHTKKNE.
Primary
IgG
bA219P18.1, chromosome 9 open reading frame 87, D4Ertd89e, FLJ10493, transmembrane protein 38BC9orf87, TRIC-BTRICB, trimeric intracellular cation channel type B
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit