Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM5 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | TMEM5 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB125189
|
Novus Biologicals
NBP310144100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
TMEM5 Polyclonal specifically detects TMEM5 in Mouse samples. It is validated for Western Blot.Specifications
TMEM5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
HP10481, putative type II membrane protein, transmembrane protein 5 | |
The immunogen is a synthetic peptide directed towards the N terminal region of mouse TMEM5 (NP_694699.1). Peptide sequence AAAAQAKERRGREQSALESEEWNPWEGDEKNEQRHRVKTNLQILNKSTKE | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
10329 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title