Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM87A Mouse anti-Human, Alexa Fluor 350, Clone: 772807, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7966U100UG
This item is not returnable.
View return policy
Description
TMEM87A Monoclonal specifically detects TMEM87A in Human samples. It is validated for Flow Cytometry.Specifications
TMEM87A | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8NBN3 | |
TMEM87A | |
E. coli-derived recombinant human TMEM87A Ser22-Asn157 Accession # Q8NBN3 | |
RUO | |
25963 | |
Human | |
IgG2b |
Flow Cytometry | |
772807 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
DKFZp564G2022, FLJ11273, MGC33727, PSEC0094, transmembrane protein 87A | |
Mouse | |
100 μg | |
Primary | |
Detects human TMEM87A in ELISAs and Western Blots | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)