Learn More
Aviva Systems Biology Corp TMEM9 Antibody - C-terminal region (ARP47253_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47253_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TMEM9. Alternate names: TMEM9AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59499