Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEPAI, Mouse anti-Human, Alexa Fluor 750, Clone: PMEPA1/2696, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279891AF750
Description
TMEPAI Monoclonal specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TMEPAI | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PMEPA1 | |
Recombinant full-length human TMEPAI protein (Uniprot: Q96W9 ) | |
0.1 mL | |
Signal Transduction | |
Human | |
IgG1 κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PMEPA1/2696 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title