Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEPAI Mouse anti-Human, Clone: rPMEPA1/6422, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP30724520UG

 View more versions of this product

Catalog No. NB121964


Add to Cart

Description

Description

TMEPAI Monoclonal specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

TMEPAI
Monoclonal
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein
Recombinant full-length human TMEPAI protein
20 μg
Signal Transduction
56937
Store at 4°C.
IgG1 κ
Immunohistochemistry (Paraffin)
rPMEPA1/6422
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit