Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEPAI Mouse anti-Human, Clone: rPMEPA1/6422, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen TMEPAI
Clone rPMEPA1/6422
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121964
View Documents
Novus Biologicals
NBP30724520UG
20 μg
Each for $159.82
Add to cart
 
NB121963
View Documents
Novus Biologicals
NBP30724510UG
100 μg
Each for $374.34
Add to cart
 
Description

Description

TMEPAI Monoclonal antibody specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

TMEPAI
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein
Recombinant full-length human TMEPAI protein
Monoclonal
Protein A or G purified
rPMEPA1/6422
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Signal Transduction
10 mM PBS with 0.05% BSA
56937
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit