Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEPAI, Mouse anti-Human, DyLight 350, Clone: PMEPA1/2696, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279891UV

Catalog No. NB011957


Add to cart

Description

Description

TMEPAI Monoclonal specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TMEPAI
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
PMEPA1
Recombinant full-length human TMEPAI protein
0.1 mL
Signal Transduction
Human
IgG1 κ
Immunohistochemistry, Immunohistochemistry (Paraffin)
PMEPA1/2696
Immunohistochemistry, Immunohistochemistry-Paraffin
prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit